Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032663.4 | NP_116052.2 | 1, 2 | 190 | ||
XM_005253962.3 | XP_005254019.1 | 1, 2 | 176 | |||
XM_005253965.4 | XP_005254022.1 | 1, 2 | 184 | |||
XM_017020048.1 | XP_016875537.1 | 1, 2 | 185 | |||
XM_017020049.1 | XP_016875538.1 | 1, 2 | 176 | |||
XM_017020053.1 | XP_016875542.1 | 1, 2 | 100 | |||
GenBank mRNA | AJ586136.1 | - | 1, 2 | 81 | ||
AK027820.1 | - | 1, 2 | 155 | |||
BC004868.2 | - | 1, 2 | 180 | 3833242 |