Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032664.3 | NP_116053.3 | 5 | 1683 | ||
XM_011544676.2 | XP_011542978.1 | 6 | 2155 | |||
XM_011544677.2 | XP_011542979.1 | 5 | 1681 | |||
XM_011544678.2 | XP_011542980.1 | 6 | 1688 | |||
XM_011544679.2 | XP_011542981.1 | 6 | 2151 | |||
XM_011544680.2 | XP_011542982.1 | 6 | 2149 | |||
XM_011544681.2 | XP_011542983.1 | 6 | 2150 | |||
GenBank mRNA | AJ431184.1 | - | 4 | 1495 | ||
AJ582015.1 | - | 5 | 1683 | |||
AK056199.1 | - | 2 | 169 | |||
AK292993.1 | - | 5 | 1681 | |||
AK297132.1 | - | 6 | 1776 | |||
BC063462.1 | - | 5 | 1723 | 5198060 |