Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRACR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144958.1 | NP_001138430.1 | 11 | 1674 | ||
NM_032680.3 | NP_116069.1 | 11 | 1674 | |||
XM_006719021.3 | XP_006719084.1 | 11 | 1674 | |||
XM_011521034.2 | XP_011519336.1 | 10 | 1421 | |||
XM_011521036.2 | XP_011519338.1 | 9 | 1181 | |||
XM_011521037.2 | XP_011519339.1 | 5 | 519 | |||
XM_011521038.2 | XP_011519340.1 | 5 | 516 | |||
GenBank mRNA | AK091124.1 | - | 5 | 510 | ||
AK304017.1 | - | 11 | 1576 | |||
BC004524.2 | - | 11 | 1571 | |||
BC150643.1 | - | 10 | 1305 | 9057013 | ||
BU632650.1 | - | 4 | 569 |