Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172677.1 | NP_001166148.1 | 5 | 2165 | ||
NM_032689.4 | NP_116078.4 | 5 | 2516 | |||
XM_006723435.3 | XP_006723498.1 | 5 | 2770 | |||
XM_006723436.2 | XP_006723499.1 | 5 | 2480 | |||
XM_006723437.2 | XP_006723500.1 | 4 | 2384 | |||
XM_006723440.1 | XP_006723503.1 | 2 | 2207 | |||
XM_011527414.2 | XP_011525716.1 | 2 | 2461 | |||
GenBank mRNA | AB209242.1 | - | 1 | 999 | ||
AK027708.1 | - | 1 | 1093 | |||
AK127464.1 | - | 1 | 1432 | |||
AK131557.1 | - | 5 | 2516 | |||
AK131559.1 | - | 5 | 2165 | |||
BC014850.1 | - | 1 | 1341 | 3447451 |