Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032701.3 | NP_116090.2 | 4 | 565 | ||
XM_005259338.3 | XP_005259395.1 | 3 | 890 | |||
XM_006723442.3 | XP_006723505.1 | 2 | 298 | |||
XM_011527415.2 | XP_011525717.1 | 4 | 1085 | |||
XM_017027388.1 | XP_016882877.1 | 4 | 538 | |||
GenBank mRNA | AF289582.1 | - | 3 | 638 | ||
AK308586.1 | - | 4 | 565 | |||
BC005842.2 | - | 2 | 299 | 2821053 | ||
BC019313.2 | - | 2 | 294 | 2821053 | ||
BC044889.1 | - | 4 | 522 | 3461265 |