Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032709.2 | NP_116098.2 | 13 | 1390 | ||
XM_011540293.2 | XP_011538595.1 | 14 | 2812 | |||
XM_011540301.2 | XP_011538603.1 | 6 | 2226 | |||
XM_017016835.1 | XP_016872324.1 | 13 | 1351 | |||
XM_017016838.1 | XP_016872327.1 | 13 | 3586 | |||
XM_017016839.1 | XP_016872328.1 | 11 | 1443 | |||
XM_017016840.1 | XP_016872329.1 | 11 | 1297 | |||
XM_017016841.1 | XP_016872330.1 | 10 | 1604 | |||
GenBank mRNA | AK022259.1 | - | 1 | 734 | ||
AK074429.1 | - | 13 | 1351 | |||
AK075265.1 | - | 13 | 1328 | |||
AK123713.1 | - | 3 | 1230 | |||
AK225637.1 | - | 13 | 1344 | |||
BC006131.1 | - | 13 | 1353 | 3626506 |