Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032726.3 | NP_116115.1 | 4, 5 | 754 | ||
XM_005246913.2 | XP_005246970.1 | 4, 5 | 716 | |||
XM_011512012.1 | XP_011510314.1 | 4, 5 | 716 | |||
XM_017005114.1 | XP_016860603.1 | 4, 5 | 657 | |||
XM_017005115.1 | XP_016860604.1 | 4, 5 | 716 | |||
XM_017005116.1 | XP_016860605.1 | 4, 5 | 716 | |||
XM_017005117.1 | XP_016860606.1 | 4, 5 | 716 | |||
XM_017005118.1 | XP_016860607.1 | 4, 5 | 1543 | |||
GenBank mRNA | AK057186.1 | - | 4, 5 | 620 | ||
AK223203.1 | - | 4, 5 | 620 | |||
AK297808.1 | - | 3, 4 | 441 | |||
AK299458.1 | - | 4, 5 | 621 | |||
AY512961.1 | - | 4, 5 | 664 | |||
BC006355.2 | - | 4, 5 | 610 | 4124286 |