Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032775.3 | NP_116164.2 | 4 | 665 | ||
NR_033825.1 | 3 | 499 | ||||
XM_017029018.1 | XP_016884507.1 | 3 | 833 | |||
XM_017029019.1 | XP_016884508.1 | 4 | 961 | |||
XM_017029020.1 | XP_016884509.1 | 5 | 2140 | |||
XM_017029021.1 | XP_016884510.1 | 4 | 594 | |||
XM_017029022.1 | XP_016884511.1 | 3 | 765 | |||
XM_017029023.1 | XP_016884512.1 | 3 | 3225 | |||
XM_017029024.1 | XP_016884513.1 | 3 | 797 | |||
XM_017029025.1 | XP_016884514.1 | 3 | 313 | |||
GenBank mRNA | AK027266.1 | - | 4 | 604 | ||
AK222850.1 | - | 4 | 571 | |||
AK290669.1 | - | 4 | 577 | |||
AK294682.1 | - | 3 | 411 | |||
BC015923.1 | - | 4 | 617 | 4562420 | ||
CR456352.1 | - | 3 | 542 |