Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032782.4 | NP_116171.3 | 3 | 684 | ||
GenBank mRNA | AB924452.1 | - | 2 | 367 | ||
AF251707.1 | - | 3 | 556 | |||
AF450242.1 | - | 3 | 487 | |||
AF450243.1 | - | 3 | 487 | |||
AK027334.1 | - | 3 | 473 | |||
AK314406.1 | - | 3 | 491 | |||
AY069944.1 | - | 3 | 430 | |||
BC063431.1 | - | 3 | 574 | 5229539 | ||
JX049979.1 | - | 3 | 430 |