Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032782.4 | NP_116171.3 | 7 | 1089 | ||
GenBank mRNA | AF251707.1 | - | 7 | 961 | ||
AF450242.1 | - | 7 | 892 | |||
AF450243.1 | - | 7 | 892 | |||
AK027334.1 | - | 7 | 878 | |||
AK130144.1 | - | 5 | 386 | |||
AK314406.1 | - | 7 | 896 | |||
AY069944.1 | - | 7 | 835 | |||
BC063431.1 | - | 7 | 979 | 5229539 | ||
JX049979.1 | - | 7 | 835 |