Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144827.1 | NP_001138299.1 | 7 | 1033 | ||
NM_032795.2 | NP_116184.2 | 7 | 1126 | |||
XM_011543039.2 | XP_011541341.1 | 7 | 1123 | |||
GenBank mRNA | AK027400.1 | - | 7 | 1080 | ||
AK225320.1 | - | 7 | 1086 | |||
AK300723.1 | - | 5 | 600 | |||
AK308059.1 | - | 7 | 986 | |||
BC014131.2 | - | 7 | 1087 | 4621434 | ||
BE260782.1 | - | 1 | 124 |