Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198696.1 | NP_001185625.1 | 9 | 1301 | ||
NM_032797.5 | NP_116186.1 | 9 | 1298 | |||
GenBank mRNA | AF337957.1 | - | 8 | 1102 | ||
AF506757.1 | - | 9 | 1298 | |||
AK027403.1 | - | 9 | 1177 | |||
AK127353.1 | - | 9 | 3053 | |||
BC006121.1 | - | 8 | 1106 | 3506309 | ||
BC023601.2 | - | 9 | 1285 | 4548458 | ||
BX537621.1 | - | 6 | 1935 |