Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032802.3 | NP_116191.2 | 14 | 1571 | ||
XM_005254722.3 | XP_005254779.1 | 15 | 1624 | |||
GenBank mRNA | AJ345028.1 | - | 14 | 1549 | ||
AJ420896.1 | - | 12 | 1122 | |||
AK027446.1 | - | 12 | 1123 | |||
AK075454.1 | - | 14 | 1560 | |||
AK315651.1 | - | 14 | 1538 | |||
AY169314.1 | - | 14 | 1567 | |||
BC025740.1 | - | 14 | 1534 | 5223715 |