Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032802.3 | NP_116191.2 | 15 | 1684 | ||
XM_005254722.3 | XP_005254779.1 | 16 | 1737 | |||
XM_017022680.1 | XP_016878169.1 | 15 | 1576 | |||
XM_017022681.1 | XP_016878170.1 | 14 | 1522 | |||
GenBank mRNA | AA988423.1 | - | 1 | 341 | ||
AJ345028.1 | - | 15 | 1662 | |||
AJ420896.1 | - | 13 | 1235 | |||
AK027446.1 | - | 13 | 1236 | |||
AK075454.1 | - | 15 | 1673 | |||
AK315651.1 | - | 15 | 1651 | |||
AY169314.1 | - | 15 | 1680 | |||
BC025740.1 | - | 15 | 1647 | 5223715 |