Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282415.1 | NP_001269344.1 | 5 | 763 | ||
NM_001282416.1 | NP_001269345.1 | 5 | 958 | |||
NM_032805.2 | NP_116194.2 | 6 | 1226 | |||
XM_017023791.1 | XP_016879280.1 | 5 | 1262 | |||
GenBank mRNA | AK027455.1 | - | 5 | 954 | ||
AK074736.1 | - | 5 | 738 | |||
BC111367.1 | - | 4 | 1555 | |||
BC114452.1 | - | 4 | 665 |