Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258452.1 | NP_001245381.1 | 14 | 2327 | ||
NM_001258453.1 | NP_001245382.1 | 15 | 2399 | |||
NM_032807.4 | NP_116196.3 | 15 | 2415 | |||
NM_178150.2 | NP_835363.1 | 14 | 2301 | |||
XM_011519748.2 | XP_011518050.1 | 16 | 2995 | |||
XM_017016842.1 | XP_016872331.1 | 15 | 3542 | |||
XM_017016843.1 | XP_016872332.1 | 16 | 2926 | |||
XM_017016844.1 | XP_016872333.1 | 14 | 2311 | |||
XM_017016845.1 | XP_016872334.1 | 14 | 2668 | |||
XM_017016846.1 | XP_016872335.1 | 13 | 2120 | |||
GenBank mRNA | AF380349.1 | - | 15 | 2415 | ||
AF454502.1 | - | 14 | 2242 | |||
AF456237.1 | - | 14 | 2257 | |||
AK027381.1 | - | 5 | 421 | |||
AK027496.1 | - | 11 | 1384 | |||
AK095343.1 | - | 14 | 2275 | |||
AK122753.1 | - | 14 | 2327 | |||
AK226128.1 | - | 14 | 2825 | |||
AK315173.1 | - | 13 | 2017 | |||
BC012762.2 | - | 1 | 43 | |||
BC020266.1 | - | 12 | 1809 | |||
BC032674.1 | - | 12 | 1753 | |||
BC080629.1 | - | 12 | 1964 | |||
BC110884.1 | - | 15 | 2324 | 6147373 | ||
BC113375.1 | - | 14 | 2248 | 8322427 | ||
BC113377.1 | - | 14 | 2248 | 8322429 |