Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321967.1 | NP_001308896.1 | 2 | 206 | ||
NM_001321968.1 | NP_001308897.1 | 2 | 206 | |||
NM_001321969.1 | NP_001308898.1 | 2 | 206 | |||
NM_032810.3 | NP_116199.2 | 2 | 578 | |||
NR_135914.1 | 2 | 206 | ||||
XM_005270252.4 | XP_005270309.1 | 2 | 300 | |||
XM_011540303.2 | XP_011538605.1 | 2 | 253 | |||
XM_017016847.1 | XP_016872336.1 | 2 | 447 | |||
XM_017016848.1 | XP_016872337.1 | 2 | 540 | |||
XM_017016849.1 | XP_016872338.1 | 2 | 329 | |||
XM_017016850.1 | XP_016872339.1 | 2 | 380 | |||
XM_017016851.1 | XP_016872340.1 | 2 | 253 | |||
GenBank mRNA | AF361493.1 | - | 1 | 412 | ||
AK075223.1 | - | 2 | 507 | |||
BC063530.1 | - | 1 | 37 | |||
BC073998.1 | - | 1 | 149 | 6199813 |