Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282736.1 | NP_001269665.1 | 13 | 2052 | ||
NM_032812.8 | NP_116201.7 | 14 | 2199 | |||
GenBank mRNA | AF378757.1 | - | 14 | 1571 | ||
AK027529.1 | - | 8 | 851 | |||
AK304591.1 | - | 15 | 2083 | |||
AY358486.1 | - | 14 | 2199 | |||
BC012885.1 | - | 13 | 1658 | 3865073 |