Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM87B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032824.2 | NP_116213.1 | 15 | 1798 | ||
XM_005263827.2 | XP_005263884.1 | 15 | 1752 | |||
XM_006712807.2 | XP_006712870.1 | 15 | 1755 | |||
XM_017005121.1 | XP_016860610.1 | 13 | 1159 | |||
GenBank mRNA | AK027587.1 | - | 15 | 1783 | ||
AK074243.1 | - | 12 | 1056 | |||
AK290294.1 | - | 15 | 1441 | |||
BC115373.1 | - | 15 | 1599 | 40026807 |