Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 15 | 1780 | ||
NM_032830.2 | NP_116219.2 | 15 | 1834 | |||
XM_005256205.3 | XP_005256262.2 | 11 | 1365 | |||
GenBank mRNA | AB075868.2 | - | 15 | 1772 | ||
AK027419.1 | - | 12 | 1389 | |||
AK027445.1 | - | 10 | 1112 | |||
AK027584.1 | - | 4 | 411 | |||
AK027634.1 | - | 15 | 1786 | |||
AK027675.1 | - | 15 | 1783 | |||
AK074795.1 | - | 15 | 1800 | |||
BC000167.2 | - | 11 | 1279 | |||
BC009348.2 | - | 15 | 1766 | 4131625 | ||
BX647265.1 | - | 15 | 1748 |