Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 8 | 972 | ||
NM_032830.2 | NP_116219.2 | 8 | 1026 | |||
XM_005256205.3 | XP_005256262.2 | 4 | 557 | |||
GenBank mRNA | AB075868.2 | - | 8 | 964 | ||
AK027419.1 | - | 5 | 581 | |||
AK027445.1 | - | 6 | 649 | |||
AK027634.1 | - | 8 | 978 | |||
AK027675.1 | - | 8 | 975 | |||
AK074795.1 | - | 8 | 992 | |||
AK310104.1 | - | 8 | 928 | |||
BC000167.2 | - | 4 | 471 | |||
BC009348.2 | - | 8 | 958 | 4131625 | ||
BX647265.1 | - | 8 | 940 |