Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032833.4 | NP_116222.4 | 1 | 1470 | ||
XM_005245551.4 | XP_005245608.2 | 1 | 1533 | |||
GenBank mRNA | AK027650.1 | - | 1 | 1470 | ||
AK222877.1 | - | 1 | 1471 | |||
AK303535.1 | - | 2 | 1024 | |||
AK316092.1 | - | 2 | 1082 | |||
AL833746.1 | - | 1 | 147 | |||
BC065280.1 | - | 1 | 1444 | 6172811 |