Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEDAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032849.3 | NP_116238.2 | 4 | 923 | ||
XM_017020801.1 | XP_016876290.1 | 3 | 1303 | |||
GenBank mRNA | AB055407.1 | - | 3 | 309 | ||
AK027740.1 | - | 522 | ||||
AK055635.1 | - | 4 | 907 | |||
BC040165.1 | - | 4 | 807 | 5590156 | ||
BC101624.1 | - | 4 | 661 | 8069130 | ||
BC101626.1 | - | 4 | 661 | 8069132 |