Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077268.1 | NP_001070736.1 | 7 | 1393 | ||
NM_001258421.1 | NP_001245350.1 | 7 | 1196 | |||
NM_032850.4 | NP_116239.3 | 8 | 1062 | |||
XM_017022683.1 | XP_016878172.1 | 6 | 1955 | |||
GenBank mRNA | AF445414.1 | - | 8 | 904 | ||
AK027746.1 | - | 7 | 825 | |||
AK122779.1 | - | 7 | 1183 | |||
AK225553.1 | - | 7 | 816 |