Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032268.4 | NP_115644.1 | 1, 2 | 1069 | ||
XM_017023793.1 | XP_016879282.1 | 1, 2 | 1043 | |||
GenBank mRNA | AF378524.1 | - | 1, 2 | 741 | ||
AK294434.1 | - | 1, 2 | 741 | |||
AL136903.1 | - | 1, 2 | 734 | |||
AL834440.1 | - | 1, 2 | 393 | |||
BC007235.1 | - | 1, 2 | 741 | 2823677 |