Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032853.3 | NP_116242.2 | 4, 5 | 325 | ||
XM_011528380.2 | XP_011526682.1 | 4, 5 | 315 | |||
XM_011528382.2 | XP_011526684.1 | 4, 5 | 315 | |||
XM_011528383.1 | XP_011526685.1 | 4, 5 | 315 | |||
XM_011528384.2 | XP_011526686.1 | 4, 5 | 315 | |||
XM_017027401.1 | XP_016882890.1 | 4, 5 | 315 | |||
GenBank mRNA | BC110874.1 | - | 3, 4 | 291 | 4933735 | |
BC144138.1 | - | 3, 4 | 241 | 9052658 | ||
BI553935.1 | - | 4, 5 | 412 | |||
BQ895780.1 | - | 4, 5 | 384 | |||
BU541563.1 | - | 4, 5 | 364 | |||
DA739708.1 | - | 4, 5 | 315 | |||
U20897.1 | - | 3, 4 | 295 |