Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSH2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291274.1 | NP_001278203.1 | 8 | 1593 | ||
NM_032855.3 | NP_116244.1 | 8 | 1060 | |||
NR_111903.1 | 8 | 1067 | ||||
XM_011528386.2 | XP_011526688.1 | 7 | 814 | |||
XM_011528387.2 | XP_011526689.1 | 7 | 816 | |||
XM_011528388.2 | XP_011526690.1 | 8 | 894 | |||
XM_011528389.2 | XP_011526691.1 | 6 | 681 | |||
XM_017027402.1 | XP_016882891.1 | 6 | 2214 | |||
XM_017027403.1 | XP_016882892.1 | 7 | 768 | |||
XM_017027404.1 | XP_016882893.1 | 6 | 652 | |||
GenBank mRNA | AK027792.1 | - | 6 | 640 | ||
AK307563.1 | - | 6 | 610 | |||
AY319652.1 | - | 8 | 1060 | |||
BC016826.1 | - | 4 | 265 | |||
BC025237.2 | - | 5 | 560 | 4853087 |