Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032856.3 | NP_116245.2 | 8 | 1157 | ||
NR_130944.1 | 7 | 1594 | ||||
NR_130945.1 | 9 | 1270 | ||||
NR_130946.1 | 8 | 1164 | ||||
NR_130947.1 | 7 | 1068 | ||||
GenBank mRNA | AF161382.1 | - | 8 | 1088 | ||
AK027200.1 | - | 3 | 1772 | |||
AK027794.1 | - | 8 | 1094 | |||
AK055578.1 | - | 7 | 1594 | |||
AK055729.1 | - | 7 | 1676 | |||
AK225068.1 | - | 8 | 1097 | |||
AK225199.1 | - | 8 | 1094 | |||
BC014115.2 | - | 8 | 1095 | |||
BC020252.2 | - | 7 | 1257 | |||
BC050648.1 | - | 8 | 1114 | |||
BC063392.1 | - | 8 | 1125 | 5168550 | ||
BM050710.1 | - | 3 | 713 |