Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032862.4 | NP_116251.4 | 1 | 168 | ||
GenBank mRNA | AK223583.1 | - | 1 | 167 |