Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128211.1 | NP_001121683.1 | 4 | 577 | ||
NM_032869.3 | NP_116258.2 | 4 | 677 | |||
XM_017013910.1 | XP_016869399.1 | 5 | 853 | |||
XM_017013911.1 | XP_016869400.1 | 5 | 623 | |||
GenBank mRNA | AF283301.1 | - | 4 | 798 | ||
AF283302.1 | - | 4 | 535 | |||
AF521133.1 | - | 4 | 577 | |||
AK027897.1 | - | 4 | 629 | |||
AK301278.1 | - | 5 | 693 | |||
AK307663.1 | - | 4 | 578 | |||
AK311583.1 | - | 5 | 629 | |||
AK315006.1 | - | 4 | 646 | |||
BC000967.2 | - | 4 | 622 | 3449287 | ||
BC031258.1 | - | 5 | 767 | 5272186 | ||
BC043406.1 | - | 4 | 642 | 6059975 |