Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032880.4 | NP_116269.3 | 6 | 1180 | ||
XM_017002604.1 | XP_016858093.1 | 6 | 826 | |||
XM_017002605.1 | XP_016858094.1 | 5 | 643 | |||
GenBank mRNA | AK075316.1 | - | 6 | 1180 | ||
AK123172.1 | - | 2 | 1121 | |||
AK294121.1 | - | 6 | 749 | |||
BC007618.1 | - | 6 | 1100 | 3355289 |