Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032880.4 | NP_116269.3 | 1, 2 | 461 | ||
XM_011542319.2 | XP_011540621.1 | 1, 2 | 418 | |||
GenBank mRNA | AK075316.1 | - | 1, 2 | 461 | ||
AK294121.1 | - | 1, 2 | 30 | |||
BC007618.1 | - | 1, 2 | 381 | 3355289 |