Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281504.1 | NP_001268433.1 | 4 | 998 | ||
NM_032885.5 | NP_116274.3 | 4 | 924 | |||
GenBank mRNA | AJ312332.1 | - | 4 | 740 | ||
AK056210.1 | - | 4 | 899 | |||
AK223383.1 | - | 4 | 849 | |||
AK294676.1 | - | 4 | 998 | |||
AK316454.1 | - | 4 | 970 | |||
BC068992.1 | - | 4 | 900 | 6578102 |