Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198836.1 | NP_001185765.1 | 3 | 2578 | ||
NM_001198837.1 | NP_001185766.1 | 2 | 2467 | |||
NM_006328.3 | NP_006319.1 | 3 | 3932 | |||
GenBank mRNA | BC007641.1 | - | 1 | 524 | ||
BC024260.1 | - | 1 | 524 | |||
CR749306.1 | - | 1 | 2137 |