Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 10 | 3301 | ||
XM_005273335.2 | XP_005273392.1 | 8 | 3142 | |||
XM_006711592.2 | XP_006711655.1 | 9 | 4089 | |||
XM_006711594.3 | XP_006711657.1 | 8 | 2797 | |||
XM_011510072.2 | XP_011508374.1 | 9 | 3141 | |||
XM_011510073.2 | XP_011508375.1 | 9 | 3619 | |||
XM_011510074.2 | XP_011508376.1 | 9 | 3160 | |||
XM_011510075.2 | XP_011508377.1 | 9 | 3647 | |||
XM_011510077.2 | XP_011508379.1 | 8 | 3083 | |||
XM_017002611.1 | XP_016858100.1 | 8 | 3557 | |||
XM_017002612.1 | XP_016858101.1 | 9 | 2966 | |||
XM_017002613.1 | XP_016858102.1 | 7 | 2926 | |||
GenBank mRNA | AK023679.1 | - | 1 | 628 | ||
AK026114.1 | - | 1 | 1191 | |||
AK056569.1 | - | 3 | 2105 | |||
AL133092.1 | - | 1 | 675 | |||
BC011542.1 | - | 1 | 1436 |