Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286490.1 | NP_001273419.1 | 15 | 1510 | ||
NM_001286491.1 | NP_001273420.1 | 15 | 1894 | |||
NM_001322949.1 | NP_001309878.1 | 14 | 1575 | |||
NM_001322950.1 | NP_001309879.1 | 14 | 1575 | |||
NM_001322951.1 | NP_001309880.1 | 13 | 1470 | |||
NM_032892.4 | NP_116281.2 | 14 | 1575 | |||
NR_104455.1 | 15 | 1891 | ||||
XM_005254730.3 | XP_005254787.1 | 14 | 1480 | |||
GenBank mRNA | AK090572.1 | - | 14 | 1507 | ||
AK123018.1 | - | 15 | 1510 | |||
AK290013.1 | - | 3 | 369 | |||
BC043486.2 | - | 15 | 1641 | |||
BC053647.1 | - | 14 | 1512 | 5578205 |