Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204300.1 | NP_001191229.1 | 7 | 1092 | ||
NM_001256423.1 | NP_001243352.1 | 8 | 1162 | |||
NM_032900.5 | NP_116289.4 | 8 | 1179 | |||
GenBank mRNA | AK074122.1 | - | 3 | 259 | ||
AK090447.1 | - | 9 | 1224 | |||
AK093316.1 | - | 6 | 810 | |||
AK093441.1 | - | 8 | 1137 | |||
AK303055.1 | - | 8 | 1140 | |||
BC113888.1 | - | 7 | 1068 | 40082328 | ||
BC114490.1 | - | 8 | 1155 | 40082327 | ||
BX648189.1 | - | 8 | 1154 | |||
CR749412.1 | - | 8 | 1155 | |||
DQ338460.1 | - | 8 | 1144 |
Gene Symbol: ARHGAP19-SLIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037909.1 | 8 | 1179 | |||
GenBank mRNA | AY336750.1 | - | 8 | 1144 |