Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286739.1 | NP_001273668.1 | 4 | 456 | ||
NM_001286740.1 | NP_001273669.1 | 3 | 321 | |||
NM_001286741.1 | NP_001273670.1 | 3 | 811 | |||
NM_001286742.1 | NP_001273671.1 | 4 | 553 | |||
NM_032907.4 | NP_116296.1 | 3 | 442 | |||
NM_201265.1 | NP_957717.1 | 3 | 371 | |||
XM_011522124.2 | XP_011520426.1 | 4 | 658 | |||
GenBank mRNA | AY037166.1 | - | 3 | 345 | ||
BC007913.2 | - | 3 | 725 | 4302762 | ||
BC030055.1 | - | 4 | 419 | 4873512 | ||
BC033919.1 | - | 4 | 460 | 4904699 | ||
BG704085.1 | - | 3 | 368 | |||
BI551787.1 | - | 3 | 428 | |||
BI552240.1 | - | 3 | 377 | |||
BP192909.1 | - | 3 | 432 | |||
HY135530.1 | - | 3 | 321 |