Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318834.1 | NP_001305763.1 | 3, 4 | 315 | ||
NM_032933.5 | NP_116322.3 | 3, 4 | 315 | |||
NR_134875.1 | 3, 4 | 315 | ||||
NR_134877.1 | 3, 4 | 315 | ||||
XM_011526233.2 | XP_011524535.1 | 3, 4 | 305 | |||
XM_017026043.1 | XP_016881532.1 | 3, 4 | 305 | |||
XM_017026044.1 | XP_016881533.1 | 3, 4 | 305 | |||
XM_017026045.1 | XP_016881534.1 | 3, 4 | 307 | |||
XM_017026047.1 | XP_016881536.1 | 3, 4 | 198 | |||
XM_017026050.1 | XP_016881539.1 | 2, 3 | 218 | |||
XM_017026051.1 | XP_016881540.1 | 3, 4 | 198 | |||
GenBank mRNA | AB253738.1 | - | 3, 4 | 215 | ||
AB253739.1 | - | 3, 4 | 215 | |||
AK126247.1 | - | 3, 4 | 307 | |||
AK303553.1 | - | 3, 4 | 199 | |||
BC114510.1 | - | 3, 4 | 251 | 40023922 | ||
BC114561.1 | - | 3, 4 | 251 | 40023923 | ||
DC334544.1 | - | 3, 4 | 317 |