Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 48, 49 | 4882 | ||
XM_006717308.2 | XP_006717371.1 | 47, 48 | 4896 | |||
XM_006717310.3 | XP_006717373.1 | 46, 47 | 2695 | |||
XM_011519138.2 | XP_011517440.1 | 48, 49 | 4514 | |||
XM_011519140.1 | XP_011517442.1 | 47, 48 | 4896 | |||
XM_011519145.2 | XP_011517447.1 | 35, 36 | 2124 | |||
GenBank mRNA | AB058773.1 | - | 45, 46 | 3369 | ||
AY149237.1 | - | 48, 49 | 4476 |