Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032583.3 | NP_115972.2 | 21 | 2889 | ||
NM_033151.3 | NP_149163.2 | 20 | 3175 | |||
NM_145186.2 | NP_660187.1 | 21 | 2889 | |||
XM_011523397.1 | XP_011521699.1 | 17 | 2395 | |||
XM_011523398.2 | XP_011521700.1 | 9 | 1110 | |||
XM_017023795.1 | XP_016879284.1 | 21 | 5503 | |||
XM_017023796.1 | XP_016879285.1 | 22 | 2976 | |||
XM_017023797.1 | XP_016879286.1 | 22 | 3104 | |||
XM_017023798.1 | XP_016879287.1 | 21 | 2930 | |||
XM_017023799.1 | XP_016879288.1 | 21 | 2912 | |||
XM_017023800.1 | XP_016879289.1 | 21 | 2903 | |||
XM_017023801.1 | XP_016879290.1 | 20 | 5395 | |||
XM_017023802.1 | XP_016879291.1 | 15 | 2250 | |||
GenBank mRNA | AF352582.1 | - | 21 | 2924 | ||
AF367202.1 | - | 21 | 2903 | |||
AF411579.1 | - | 21 | 2903 | |||
AK301547.1 | - | 20 | 2833 | |||
AL117406.1 | - | 1 | 82 | |||
AY040219.1 | - | 20 | 3175 |