Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033089.6 | NP_149080.2 | 1 | 885 | ||
GenBank mRNA | AK025768.1 | - | 2 | 170 | ||
BC069238.1 | - | 1 | 885 | 6287290 | ||
BC107584.1 | - | 2 | 387 | 6457775 |