Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033118.3 | NP_149109.1 | 11 | 1649 | ||
GenBank mRNA | AF325549.1 | - | 10 | 1547 | ||
AJ272502.1 | - | 11 | 1655 | |||
BC007753.1 | - | 4 | 485 | |||
BC069627.1 | - | 10 | 1547 | 7262446 | ||
BC092413.1 | - | 11 | 1649 | 30530456 | ||
BC127622.1 | - | 10 | 1547 | 40085085 | ||
BX647635.1 | - | 11 | 3785 |