Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033386.3 | NP_203744.1 | 2 | 449 | ||
XM_005261791.3 | XP_005261848.1 | 2 | 262 | |||
XM_005261792.3 | XP_005261849.1 | 2 | 262 | |||
XM_011530471.2 | XP_011528773.1 | 2 | 262 | |||
XM_011530472.2 | XP_011528774.1 | 2 | 262 | |||
XM_011530473.2 | XP_011528775.1 | 2 | 262 | |||
XM_011530474.2 | XP_011528776.1 | 2 | 673 | |||
XM_011530475.2 | XP_011528777.1 | 2 | 262 | |||
XM_011530476.2 | XP_011528778.1 | 2 | 262 | |||
XM_017029026.1 | XP_016884515.1 | 2 | 421 | |||
GenBank mRNA | AJ496196.1 | - | 2 | 241 | ||
AL834373.1 | - | 2 | 103 | |||
CB997694.1 | - | 2 | 327 | |||
CR456437.1 | - | 2 | 180 | |||
DR004306.1 | - | 2 | 431 |