Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102450.2 | NP_001095920.1 | 6 | 576 | ||
NM_033345.3 | NP_203131.1 | 5 | 447 | |||
XM_005245555.3 | XP_005245612.1 | 6 | 590 | |||
XM_011510089.2 | XP_011508391.1 | 6 | 1487 | |||
XM_011510090.2 | XP_011508392.1 | 9 | 1317 | |||
XM_011510091.2 | XP_011508393.1 | 5 | 464 | |||
XM_017002631.1 | XP_016858120.1 | 9 | 937 | |||
XM_017002632.1 | XP_016858121.1 | 7 | 768 | |||
XM_017002633.1 | XP_016858122.1 | 6 | 966 | |||
XM_017002634.1 | XP_016858123.1 | 6 | 645 | |||
XM_017002635.1 | XP_016858124.1 | 7 | 1092 | |||
XM_017002636.1 | XP_016858125.1 | 5 | 574 | |||
XM_017002637.1 | XP_016858126.1 | 6 | 772 | |||
GenBank mRNA | AF300649.1 | - | 4 | 285 | ||
AI209102.1 | - | 2 | 337 | |||
AK294660.1 | - | 6 | 543 | |||
AK296514.1 | - | 6 | 536 | |||
BC069677.1 | - | 6 | 525 | 7262579 | ||
BC103865.1 | - | 6 | 552 | 40003386 | ||
BC103866.1 | - | 6 | 552 | 40003387 | ||
BC103867.1 | - | 6 | 552 | 40003388 | ||
BX281622.1 | - | 5 | 458 | |||
BX954018.1 | - | 5 | 351 |