Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032364.5 | NP_115740.5 | 2 | 1412 | ||
GenBank mRNA | AK055945.1 | - | 1 | 450 | ||
AK123025.1 | - | 1 | 992 | |||
AK291222.1 | - | 2 | 1647 | |||
AY188447.1 | - | 2 | 1329 | |||
BC080655.1 | - | 1 | 364 | |||
BC117146.1 | - | 2 | 1283 | 40125697 | ||
BC117148.1 | - | 2 | 1283 | 40125699 | ||
EF560741.1 | - | 2 | 1334 |