Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033114.3 | NP_149105.3 | 2, 3 | 283 | ||
GenBank mRNA | AB062361.1 | - | 2, 3 | 283 | ||
AK308244.1 | - | 2, 3 | 153 | |||
BC005099.1 | - | 2, 3 | 233 | |||
BC010177.2 | - | 2, 3 | 248 | |||
BC022543.1 | - | 2, 3 | 258 | 4797962 |