Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033122.3 | NP_149113.3 | 1 | 618 | ||
GenBank mRNA | AF380838.1 | - | 1 | 588 | ||
AK057459.1 | - | 1 | 591 | |||
AK315022.1 | - | 1 | 591 | |||
BC046111.1 | - | 1 | 580 | 5171504 |