Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033105.4 | NP_149096.2 | 4 | 515 | ||
XM_011517618.2 | XP_011515920.1 | 4 | 447 | |||
XM_011517619.2 | XP_011515921.1 | 2 | 259 | |||
XM_011517620.2 | XP_011515922.1 | 4 | 508 | |||
XM_011517621.2 | XP_011515923.1 | 2 | 205 | |||
GenBank mRNA | AF368276.1 | - | 4 | 493 | ||
AL133660.1 | - | 5 | 2001 | |||
BC016742.2 | - | 4 | 512 | 4830917 | ||
BG701336.1 | - | 4 | 533 |