Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042717.2 | NP_001036182.1 | 8 | 832 | ||
NM_033107.3 | NP_149098.2 | 10 | 1069 | |||
GenBank mRNA | AF351613.1 | - | 8 | 833 | ||
AK095561.1 | - | 10 | 1059 | |||
AK125553.1 | - | 1 | 66 | |||
AK294325.1 | - | 8 | 782 | |||
BC004923.2 | - | 9 | 966 | |||
BC021573.1 | - | 9 | 966 | |||
BC107714.1 | - | 10 | 1053 | 6714887 | ||
BC144162.1 | - | 8 | 778 | |||
CR933597.1 | - | 9 | 883 |